Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M 344, Tocris Bioscience™

Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
251456-60-7
C16H25N3O3
307.394
MXWDSZWTBOCWBK-UHFFFAOYSA-N
4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl benzamide, histone deacetylase inhibitor iii, n-hydroxy-7-4-dimethylaminobenzoyl aminoheptanamide, 4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl-benzamide, 4-dimethylamino-n-6-hydroxycarbamoylhexyl benzamide, n-hydroxy-7-4-dimethylaminobenzoyl-aminoheptanamide, 4-dimethylamino-n-6-hydroxycarbamoylhexyl-benzamide, benzamide, 4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl, 7-4-dimethylamino phenyl formamido-n-hydroxyheptanamide
3994
4-(dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide
CN(C)C1=CC=C(C=C1)C(=O)NCCCCCCC(=O)NO
This item is not returnable. View return policy
251456-60-7
C16H25N3O3
307.394
MXWDSZWTBOCWBK-UHFFFAOYSA-N
4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl benzamide, histone deacetylase inhibitor iii, n-hydroxy-7-4-dimethylaminobenzoyl aminoheptanamide, 4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl-benzamide, 4-dimethylamino-n-6-hydroxycarbamoylhexyl benzamide, n-hydroxy-7-4-dimethylaminobenzoyl-aminoheptanamide, 4-dimethylamino-n-6-hydroxycarbamoylhexyl-benzamide, benzamide, 4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl, 7-4-dimethylamino phenyl formamido-n-hydroxyheptanamide
3994
4-(dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide
CN(C)C1=CC=C(C=C1)C(=O)NCCCCCCC(=O)NO

Histone deacetylase inhibitor

Histone deacetylase inhibitor (IC50 = 100 nM). Induces terminal cell differentiation and causes an increase in hyperacetylated histone H4. Antiproliferative agent; suppresses the growth of human endometrial and ovarian cancer cells by inducing cell cycle arrest and apoptosis.
Quantity 50 mg
Formula Weight 307.39
Percent Purity >98%
Chemical Name or Material M 344

Recommended Storage : Store at Room Temperature