Learn More
BUCHI M-565 Melting Point Instrument with Sample Loader M-569: PROMO
Automatic and reliable determination of melting point, boiling point and slip melting point.
Manufacturer:  BUCHI 051998PM
Catalog No. 05-422-993PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Sample loader M-569, Calibration set M-565, melting point tubes (100 pieces), boiling point tubes (10 pieces), boiling capillary (10 pieces), sample holder, cleaning tool, CD demo melting point monitor, protection cover, USB cable (2m), operation manu
Description
Description
Automatic and reliable determination of melting point, boiling point and slip melting point.
Specifications
Specifications
Melting Point Instrument
37cm
Determining Melting Point, Boiling Point and Sling Melting Point
20cm
M-565
10° to 400°C
19cm
4.5kg
14.5 in.
TFT
7.8 in.
Sample loader M-569, Calibration set M-565, melting point tubes (100 pieces), boiling point tubes (10 pieces), boiling capillary (10 pieces), sample holder, cleaning tool, CD demo melting point monitor, protection cover, USB cable (2m), operation manual
0.1°C (temp.)
7.4 in.
100-240V 50/60Hz 150w