Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Anisic hydrazide, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
5785-06-8
C8H10N2O2
CH3OC6H4CONHNH
MFCD00007601
3-methoxybenzhydrazide,m-anisic hydrazide,m-anisohydrazide,m-methoxy benzhydrazide,3-methoxybenzoic hydrazide,3-methoxybenzenecarbohydrazide,3-methoxybenzene-1-carbohydrazide,benzoic acid, 3-methoxy-, hydrazide,m-anisoylhydrazine,m-methoxybenzohydrazide
m-Anisic hydrazide
5785-06-8
C8H10N2O2
CH3OC6H4CONHNH
MFCD00007601
3-methoxybenzhydrazide,m-anisic hydrazide,m-anisohydrazide,m-methoxy benzhydrazide,3-methoxybenzoic hydrazide,3-methoxybenzenecarbohydrazide,3-methoxybenzene-1-carbohydrazide,benzoic acid, 3-methoxy-, hydrazide,m-anisoylhydrazine,m-methoxybenzohydrazide
m-Anisic hydrazide
Quantity 5g
Packaging Glass bottle
InChI Key VMZSDAQEWPNOIB-UHFFFAOYSA-N
Molecular Weight (g/mol) 166.18
PubChem CID 79848
Formula Weight 166.18

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.