Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ m-Anisidine, 98%

Manufacturer:  Alfa Aesar™ A1081530

Quantity 250g

 View more versions of this product

Catalog No. AAA1081530


  Check Availability
Add to cart

Specifications

Description

Applications
The unusually large amount of dibromo product produced upon bromination of m-anisidine may be attributed to the two doubly activeated positions. The best enantioselectivity of 97 % ee was observed for the reaction of m-anisidine in organocatalytic asymmetric three-component cyclization of cinnamaldehydes and primary amines with 1, 3-Dicarbonyl Compounds. Evidence for the control of 2nd-harmonic generation activities from the x-ray crystal-structures of the complexes of l-tartaric acid with m-anisidine and p-toluidine was determined.

Solubility
Soluble in water(18g/L).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

536-90-3
123.16
UN2431
14,667
m-Anisidine
C7H9NO
MFCD00007783
386119
3-Aminoanisole; 3-Methoxyaniline

Specifications

126°C (258°F)
250° to 251°C
250g
C7H9NO
MFCD00007783
386119
3-Aminoanisole; 3-Methoxyaniline
98%
Characteristic
−1°C
536-90-3
123.16
UN2431
14,667
m-Anisidine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H311-H373-H302-H332
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P280h-P273-P309-P310-P302+P352-P501a
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANISIDINES

RTECSNumber : BZ5408000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit