Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
m-Anisoyl chloride, 99%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Specifications
Flash Point | 92°C |
Specific Gravity | 1.214 |
Boiling Point | 123°C to 125°C (15.0mmHg) |
Refractive Index | 1.5575 to 1.5595 |
Density | 1.2140g/mL |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Color | Colorless to Yellow |
Quantity | 100g |
Assay Percent Range | 99% |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 216-975-2