Learn More
Zyagen LabsSupplier Diversity Partner Mouse CD1 mammary E18 frozen sections are made of normal healthy freshly harvested mammary gland of CD1 Mouse. Sections of 10 microns are mounted on positively charged slides. Ideal for immunohistochemistry & in situ hybridization. 10 slides/PK. DFS Item
Manufacturer:  Zyagen LabsSupplier Diversity Partner MF41418
Zyagen Frozen tissue sections are made of normal healthy tissues which are freshly harvested of single donor and snapped frozen in OCT blocks without any fixative. Blocks are sectioned by cryostat at a thickness of 7 or 10microns and sections mounted on positively charged slides (2 sections per slide). Frozen tissue sections are prepared in RNase- and protease-free environment and are ideal for immunohistochemistry and in situ hybridization. Tissue sections are sold in a unit of 10 slides. Frozen sections are shipped in vacuum sealed slide boxes or slide mailers on dry ice and are good for one year if properly stored at-80oC. Quality of tissue morphology is tested using H&E staining.
Catalog No. 50-172-0084