Learn More
m-Cresol Purple, Thermo Scientific Chemicals
Description
m-Cresol Purple acts as a pH indicator dye. It plays an important role in spectrophotometric pH measurements. It operates in the pH ranges 1.2 - 2.8 (red to yellow) and 7.4 - 9.0 (yellow to violet). Further, it is employed to stain epithelial cells and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
m-Cresol Purple acts as a pH indicator dye. It plays an important role in spectrophotometric pH measurements. It operates in the pH ranges 1.2 - 2.8 (red to yellow) and 7.4 - 9.0 (yellow to violet). Further, it is employed to stain epithelial cells and proteins.
Solubility
Soluble in water and sodium hydroxide solution. Slightly soluble in alcohol.
Notes
Transition interval (acid): pH 1.2 (red) to pH 2.8 (yellow); (alkaline) pH 7.4 (yellow) to pH 9.0 (purple)Incompatible with strong oxidizing agents.
Specifications
Beilstein | 350314 |
Merck Index | 14,2577 |
Solubility Information | Soluble in water and sodium hydroxide solution. Slightly soluble in alcohol. |
Formula Weight | 382.44 |
Quantity | 5 g |
Chemical Name or Material | m-Cresol Purple |
Safety and Handling
EINECSNumber | 218-960-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.