Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol Purple, sodium salt, pure, water soluble, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
Packaging:
Glass bottle
62625-31-4
C21H17NaO5S
MFCD00010177
m-Cresol Purple, sodium salt
62625-31-4
C21H17NaO5S
MFCD00010177
m-Cresol Purple, sodium salt
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 404.4
Physical Form Powder
Grade Pure
Packaging Glass bottle
Solubility Solubility in water: soluble.
Visual Transition Interval from pH 7.4 (yellow) to pH 9.0 (purple)
Color Black to Brown-Orange
Quantity 50g
Chemical Name or Material m-Cresol Purple, sodium salt
Show More Show Less

EINECSNumber : 263-656-9

TSCA : TSCA