Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol Purple, T.S., BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

High quality chemicals for laboratory and specialized industrial use

CAS 7732-18-5
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.03kg
Quantity 500mL
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material m-Cresol Purple, T.S.