Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M-CSF R/CD115 Mouse anti-Human, Biotin, Clone: 6B9H2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237289B

Catalog No. NBP237289B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

M-CSF R/CD115
6B9H2
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CSF1R
Purified recombinant fragment of human CSF1R (AA: 344-497) expressed in E. Coli.
Protein A or G purified
Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase
Primary
1436
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
PBS with 0.05% Sodium Azide
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit