Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
M-CSFR/CD115 Rabbit anti-Human, Clone: 29, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | M-CSFR/CD115 |
---|---|
Clone | 29 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Human samples. It is validated for ELISA.Specifications
M-CSFR/CD115 | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms | |
Recombinant Human M-CSFR/CD115 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
29 | |
ELISA | |
Unconjugated | |
Rabbit | |
Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase | |
0.2 um filtered solution in PBS | |
1436 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title