Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Ethyltoluene, 98%, Acros Organics

Click to view available options
Quantity:
10g
25g
620-14-4
C9H12
120.195
ZLCSFXXPPANWQY-UHFFFAOYSA-N
3-ethyltoluene,m-ethyltoluene,1-methyl-3-ethylbenzene,benzene, 1-ethyl-3-methyl,m-ethylmethylbenzene,m-methylethylbenzene,toluene, m-ethyl,3-methylethylbenzene,1-ethyl-3-methyl-benzene,unii-737ptd7o7e
12100
CHEBI:77512
1-ethyl-3-methylbenzene
CCC1=CC=CC(=C1)C
620-14-4
C9H12
120.195
ZLCSFXXPPANWQY-UHFFFAOYSA-N
3-ethyltoluene,m-ethyltoluene,1-methyl-3-ethylbenzene,benzene, 1-ethyl-3-methyl,m-ethylmethylbenzene,m-methylethylbenzene,toluene, m-ethyl,3-methylethylbenzene,1-ethyl-3-methyl-benzene,unii-737ptd7o7e
12100
CHEBI:77512
1-ethyl-3-methylbenzene
CCC1=CC=CC(=C1)C
Boiling Point 158.0°C to 159.0°C
Refractive Index 1.4956
Density 0.8600g/mL
Melting Point -97.0°C to +95.0°C
Quantity 10g
Flash Point 38°C
Specific Gravity 0.86
Assay Percent Range 98%
Linear Formula C2H5C6H4CH3
Formula Weight 120.19
Percent Purity 98%
Chemical Name or Material m-Ethyltoluene
Show More Show Less