Learn More
GVS™ M-Green Yeast and Mold Bottled Agar
Ready-to-use 100 mL bottled M-Green Yeast and Mold Agar liquid culture growth media can save time and money by shortening prep time and minimizing possible contamination.
Manufacturer:  GVS™ 10496705
Catalog No. 10-486-159
Add to cart
Description
Description
Description
  • Used for detecting fungi in routine analysis of beverages using membrane filter technique.
  • The addition of bromocresol green, which diffuses into fungal colonies as an alkaline reaction, allows them to be easily identified.
  • Metabolic by-products from the developing colonies diffuse into the surrounding medium, further reducing the pH which aids in the inhibition of bacterial growth, but also produces an acid reaction that causes residual bromocresol green to change to yellow.
Features and Benefits
  • Pre-Sterilized and Ready to Use
  • Time Saving, 100 mL bottled solution, Reduce Risk of Cross-Contamination.
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability
Applications
  • Microbiological analysis.
  • Used for detecting fungi in routine analysis of beverages using membrane filter technique.
  • For testing soft drinks and other beverages for growth of yeast and mold
Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits.
This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.
See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.
Specifications
Specifications
1
M-Green Yeast and Mold Bottled Agar
Agar
100 mL
Bottle
yeast and mold in beverages and food.
US
Batch-specific quality certificate in each pack