Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Hydroxycinnamic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
14755-02-3
C9H8O3
164.16
MFCD00004390
KKSDGJDHHZEWEP-SNAWJCMRSA-N
3-hydroxycinnamic acid,3-coumaric acid,m-coumaric acid,m-hydroxycinnamic acid,3-3-hydroxyphenyl acrylic acid,e-3-3-hydroxyphenyl acrylic acid,trans-3-hydroxycinnamic acid,trans-3-hydroxycinnamate,cinnamic acid, m-hydroxy,2e-3-3-hydroxyphenyl prop-2-enoic acid
637541
CHEBI:32357
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC(=C1)O)C=CC(=O)O
14755-02-3
C9H8O3
164.16
MFCD00004390
KKSDGJDHHZEWEP-SNAWJCMRSA-N
3-hydroxycinnamic acid,3-coumaric acid,m-coumaric acid,m-hydroxycinnamic acid,3-3-hydroxyphenyl acrylic acid,e-3-3-hydroxyphenyl acrylic acid,trans-3-hydroxycinnamic acid,trans-3-hydroxycinnamate,cinnamic acid, m-hydroxy,2e-3-3-hydroxyphenyl prop-2-enoic acid
637541
CHEBI:32357
(E)-3-(3-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC(=C1)O)C=CC(=O)O
Packaging Glass Bottle
Melting Point 189°C to 192°C
Quantity 25g
Infrared Spectrum Authentic
Color Beige
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC6H4CH=CHCO2H
Beilstein 10, 294
Formula Weight 164.16
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material m-Hydroxycinnamic acid, Predominantly trans
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning