Learn More
Brady™ M Series MetaLabel™ Solvent Resistant Metallized Polyester Labels, Black on Light Gray
Metallized Polyester (B-428) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
$66.80 - $81.10
Specifications
Product Type Label
Print Technology Thermal Transfer
Quantity Cartridge of 260 Labels
Labels per Row 1
For Use With (Equipment) BMP™51 Label Maker; BMP™53 Label Maker
View More Specs
Products 2
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
19-000-585 View Documents Brady™
M30428
1.5H x 0.75 in. W Each for $66.80
Add to cart
19-000-584 View Documents Brady™
M29428
1.5H x 0.5 in. W Each for $81.10
Add to cart
Description
Description
  • Matte, light gray finish gives labels a rating and serial plate appearance
  • Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • UL approved
  • CSA accepted
  • All-in-one cartridge includes label supply and R4300 Series ribbon
Specifications
Specifications
Label
Cartridge of 260 Labels
BMP™51 Label Maker; BMP™53 Label Maker
Light Gray
ASTM D 1000; ASTM D 2979; ASTM G 155; CSA; Halogen Free (DIN VDE 0472 Part 815); UL Recognized
Thermal Transfer
1
Metallized Polyester
Permanent Acrylic