Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Terphenyl 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
92-06-8
C18H14
230.31
MFCD00003059
YJTKZCDBKVTVBY-UHFFFAOYSA-N
m-terphenyl,1,1':3',1-terphenyl,m-diphenylbenzene,m-triphenyl,santowax m,isodiphenylbenzene,1,3-terphenyl,meta-terphenyl,santowax om,1,1'-biphenyl, 3-phenyl
7076
1,3-diphenylbenzene
C1=CC=C(C=C1)C2=CC(=CC=C2)C3=CC=CC=C3
92-06-8
C18H14
230.31
MFCD00003059
YJTKZCDBKVTVBY-UHFFFAOYSA-N
m-terphenyl,1,1':3',1-terphenyl,m-diphenylbenzene,m-triphenyl,santowax m,isodiphenylbenzene,1,3-terphenyl,meta-terphenyl,santowax om,1,1'-biphenyl, 3-phenyl
7076
1,3-diphenylbenzene
C1=CC=C(C=C1)C2=CC(=CC=C2)C3=CC=CC=C3
Color White-Yellow
Quantity 25g
Melting Point 86°C
Boiling Point 365°C
UN Number 3077
Formula Weight 230.31
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material m-Terphenyl

EINECSNumber : (4)-0017

RTECSNumber : WZ6470000

TSCA : Yes