Alfa Aesar™ m-Thiocresol, 97%

Manufacturer: Alfa Aesar™ B2381822

Quantity 100g

 View more versions of this product

Catalog No. AAB2381822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-40-7
    Molecular Formula C7H8S
    Formula Weight 124.21
    MDL Number MFCD00004843
    UN Number UN2810
    Beilstein 1851959
    Merck Index 14,9325
    Synonym 3-Mercaptotoluene; 3-Methylthiophenol
    Chemical Name or Material m-Thiocresol

    Specifications

    Formula Weight 124.21
    MDL Number MFCD00004843
    UN Number UN2810
    Merck Index 14,9325
    Synonym 3-Mercaptotoluene; 3-Methylthiophenol
    Health Hazard 1 Danger
    Boiling Point 196°C
    Melting Point <−20°C
    Flash Point 72°C (161°F)
    Quantity 100g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    RTECS Number XT8700000
    TSCA TSCA
    Molecular Formula C7H8S
    CAS 108-40-7
    Beilstein 1851959
    Health Hazard 2 GHS H Statement
    H331-H318-H227-H302-H312-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material m-Thiocresol
    Assay 97%

    GHS H Statement
    H331-H318-H227-H302-H312-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit