Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ m-Toluoyl chloride, 99%

Click to view available options
Quantity:
25g
100g
500g
1711-06-4
C8H7ClO
154.593
MFCD00000681
YHOYYHYBFSYOSQ-UHFFFAOYSA-N
m-toluoyl chloride,m-methylbenzoyl chloride,3-toluoyl chloride,toluoyl chloride,benzoyl chloride, 3-methyl,m-toluyl chloride,meta methyl benzoyl chloride,3-methylbenzoylchloride,3-toluoylchloride,unii-w2vjw4350k
74375
3-methylbenzoyl chloride
CC1=CC=CC(=C1)C(=O)Cl
1711-06-4
C8H7ClO
154.593
MFCD00000681
YHOYYHYBFSYOSQ-UHFFFAOYSA-N
m-toluoyl chloride,m-methylbenzoyl chloride,3-toluoyl chloride,toluoyl chloride,benzoyl chloride, 3-methyl,m-toluyl chloride,meta methyl benzoyl chloride,3-methylbenzoylchloride,3-toluoylchloride,unii-w2vjw4350k
74375
3-methylbenzoyl chloride
CC1=CC=CC(=C1)C(=O)Cl
Applications
m-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, active metals, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Odor Pungent
Quantity 100g
Flash Point 76°C (168°F)
Boiling Point 220°C
Melting Point -23°C
UN Number UN3265
Beilstein 878419
Formula Weight 154.6
Percent Purity 99%
Chemical Name or Material m-Toluoyl chloride

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : DM6644000

TSCA : TSCA