Alfa Aesar™ m-Toluoyl chloride, 99%

Manufacturer: Alfa Aesar™ B2391436

Quantity 500g

 View more versions of this product

Catalog No. AAB2391436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1711-06-4
    Molecular Formula C8H7ClO
    Formula Weight 154.6
    MDL Number MFCD00000681
    UN Number UN3265
    Beilstein 878419
    Synonym 3-Methylbenzoyl chloride
    Chemical Name or Material m-Toluoyl chloride

    Specifications

    Formula Weight 154.6
    Flash Point 76°C (168°F)
    Quantity 500g
    TSCA TSCA
    Molecular Formula C8H7ClO
    MDL Number MFCD00000681
    CAS 1711-06-4
    Odor Pungent
    UN Number UN3265
    Synonym 3-Methylbenzoyl chloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Boiling Point 220°C
    Melting Point −23°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    RTECS Number DM6644000
    Beilstein 878419
    Chemical Name or Material m-Toluoyl chloride
    Assay 99%

    Description

    Applications
    m-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.

    Solubility
    Reacts with water.

    Notes
    Moisture sensitive. Store away from oxidizing agents, bases, active metals, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit