Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
m-Tolyl acetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Density | 1.05 |
Boiling Point | 100°C (14 mmHg) |
Flash Point | 95°C (203°F) |
Refractive Index | 1.502 |
Quantity | 25 g |
Beilstein | 1862796 |
Merck Index | 14,2583 |
Solubility Information | Not miscible or difficult to mix with water. |
Formula Weight | 150.18 |
Percent Purity | 97% |
Show More |
Safety and Handling
EINECSNumber | 204-546-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction