Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Tolylhydrazine Hydrochloride 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
637-04-7
C7H11ClN2
158.629
MFCD00012932
SHTBTGXNVWYODU-UHFFFAOYSA-N
3-methylphenylhydrazine hydrochloride,m-tolylhydrazine hydrochloride,m-tolylhydrazinium 1+ chloride,3-methylphenyl hydrazine hydrochloride,3-methylphenylhydrazine hcl,m-tolyhydrazine hydrochloride,1-3-methylphenyl hydrazine hydrochloride,3-tolylhydrazine hydrochloride,3-methyl phenylhydrazine hydrochloride,hydrazine, 3-methylphenyl-, monohydrochloride
71562
(3-methylphenyl)hydrazine;hydrochloride
CC1=CC(=CC=C1)NN.Cl
637-04-7
C7H11ClN2
158.629
MFCD00012932
SHTBTGXNVWYODU-UHFFFAOYSA-N
3-methylphenylhydrazine hydrochloride,m-tolylhydrazine hydrochloride,m-tolylhydrazinium 1+ chloride,3-methylphenyl hydrazine hydrochloride,3-methylphenylhydrazine hcl,m-tolyhydrazine hydrochloride,1-3-methylphenyl hydrazine hydrochloride,3-tolylhydrazine hydrochloride,3-methyl phenylhydrazine hydrochloride,hydrazine, 3-methylphenyl-, monohydrochloride
71562
(3-methylphenyl)hydrazine;hydrochloride
CC1=CC(=CC=C1)NN.Cl
Quantity 25g
Color Undesignated
Formula Weight 158.63
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material m-Tolylhydrazine Hydrochloride

EINECSNumber : (3)-0468

TSCA : No