Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Tolylmagnesium chloride, 1M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
121905-60-0,109-99-9
C7H7ClMg
150.89
YIDAONIIDXWYSC-UHFFFAOYSA-M
m-tolylmagnesium chloride,m-tolyl magnesium chloride,m-methylphenylmagnesium chloride,bvycqpclcztspd-uhfffaoysa-m,m-tolylmagnesium chloride solution 1.0 m in thf,m-tolylmagnesium chloride solution, 1.0 m in thf
4384483
magnesium;methylbenzene;chloride
CC1=C[C-]=CC=C1.[Mg+2].[Cl-]
121905-60-0,109-99-9
C7H7ClMg
150.89
YIDAONIIDXWYSC-UHFFFAOYSA-M
m-tolylmagnesium chloride,m-tolyl magnesium chloride,m-methylphenylmagnesium chloride,bvycqpclcztspd-uhfffaoysa-m,m-tolylmagnesium chloride solution 1.0 m in thf,m-tolylmagnesium chloride solution, 1.0 m in thf
4384483
magnesium;methylbenzene;chloride
CC1=C[C-]=CC=C1.[Mg+2].[Cl-]
CAS Min % 14.99,82.64
CAS Max % 17.36,85.01
Solubility Information Solubility in water: reacts
Formula Weight 150.89
Physical Form Solution
Concentration 0.95 to 1.10M
Density 0.9560g/mL
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: reacts.
Specific Gravity 0.956
Color Undesignated
Quantity 800mL
Flash Point −21°C
Chemical Name or Material m-Tolylmagnesium chloride
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger