Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Argos Technologies M-Vac Aspirating System

Argos M-Vac and M-Vac Jr. systems are used to collect a variety of liquids and supernatants including tissue and cell culture media, biological immunology reagents, or any liquid chemical waste that uses a standard in-house vacuum. 

Manufacturer:  Argos Technologies EV612

 View more versions of this product

Catalog No. 03-391-215


Add to cart

Description

Description

Ideal for use with critical or dangerous liquid Autoclavable at 121°C for 20 minutes Withstands 700mm Hg
Specifications

Specifications

Red
Barbed tube fittings
1
M-Vac Jr. 2L Polypropylene bottle
Red
M-Vac Jr. Lid with barbed tube fittings for 2L bottle 1/EA
Polypropylene (PP)
Bottle closure
Yes
53B
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit