Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylene, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
108-38-3
C8H10
106.168
MFCD00008536
IVSZLXZYQVIEFR-UHFFFAOYSA-N
m-xylene,1,3-dimethylbenzene,m-xylol,m-dimethylbenzene,meta-xylene,m-methyltoluene,3-xylene,benzene, 1,3-dimethyl,1,3-dimethylbenzol,santosol 150
7929
CHEBI:28488
1,3-xylene
CC1=CC(=CC=C1)C
108-38-3
C8H10
106.168
MFCD00008536
IVSZLXZYQVIEFR-UHFFFAOYSA-N
m-xylene,1,3-dimethylbenzene,m-xylol,m-dimethylbenzene,meta-xylene,m-methyltoluene,3-xylene,benzene, 1,3-dimethyl,1,3-dimethylbenzol,santosol 150
7929
CHEBI:28488
1,3-xylene
CC1=CC(=CC=C1)C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C6H4(CH3)2
Beilstein 05, 370
Merck Index 15, 10278
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Formula Weight 106.17
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Density 0.8640g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980
Residue after Evaporation 0.005% max.
Solubility Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, ether and most organic, solvents
Specific Gravity 0.864
Water 0.1% max.
Flash Point 25°C
Viscosity 0.62 mPa.s (20°C)
Boiling Point 139.0°C
Color Colorless
Melting Point -48.0°C
Quantity 250mL
Chemical Name or Material m-Xylene
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning