Learn More
m-Xylylene dichloride, 97%, Thermo Scientific Chemicals
Description
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α?-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | 33°C to 37°C |
Density | 1.302 |
Boiling Point | 250°C to 255°C |
Flash Point | 144°C (291°F) |
Quantity | 25 g |
UN Number | UN2928 |
Beilstein | 2079898 |
Solubility Information | Insoluble in water. |
Formula Weight | 175.06 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | H302-H314-H330 |
Hazard Statement | GHS H Statement H330-H314-H318-H302 Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
Precautionary Statement | P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S. |
EINECSNumber | 210-932-1 |
RTECSNumber | ZE4070000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.