Alfa Aesar™ m-Xylylene dichloride, 97%

Manufacturer: Alfa Aesar™ A1154809

Quantity 10g

 View more versions of this product

Catalog No. AAA1154809

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 626-16-4
    Molecular Formula C8H8Cl2
    Formula Weight 175.06
    MDL Number MFCD00000912
    UN Number UN2928
    Beilstein 2079898
    Synonym 1,3-Bis(chloromethyl)benzene; alpha,alpha'-Dichloro-m-xylene
    Chemical Name or Material m-Xylylene dichloride

    Specifications

    Formula Weight 175.06
    Health Hazard 3 GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H8Cl2
    CAS 626-16-4
    RTECS Number ZE4070000
    Synonym 1,3-Bis(chloromethyl)benzene; alpha,alpha'-Dichloro-m-xylene
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
    TSCA TSCA
    UN Number UN2928
    Beilstein 2079898
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H330-H314-H318-H302
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Boiling Point 250° to 255°C
    Melting Point 33° to 37°C
    Flash Point 144°C (291°F)
    Quantity 10g
    MDL Number MFCD00000912
    Chemical Name or Material m-Xylylene dichloride
    Assay 97%

    Description

    Applications
    m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α ft.-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

    Solubility
    Insoluble in water.

    Notes
    Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

    GHS H Statement
    H330-H314-H318-H302
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P273-P305+P351+P338-P309-P310-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit