Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Xylylenediamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
Packaging:
Glass bottle
1477-55-0
C8H12N2
136.2
MFCD00008119
FDLQZKYLHJJBHD-UHFFFAOYSA-N
m-xylylenediamine,1,3-benzenedimethanamine,1,3-bis aminomethyl benzene,1,3-xylylenediamine,1,3-phenylenedimethanamine,m-xylylendiamin,1,3-xylenediamine,mxda,m-phenylenebis methylamine,methylamine, m-phenylenebis
15133
[3-(aminomethyl)phenyl]methanamine
C1=CC(=CC(=C1)CN)CN
1477-55-0
C8H12N2
136.2
MFCD00008119
FDLQZKYLHJJBHD-UHFFFAOYSA-N
m-xylylenediamine,1,3-benzenedimethanamine,1,3-bis aminomethyl benzene,1,3-xylylenediamine,1,3-phenylenedimethanamine,m-xylylendiamin,1,3-xylenediamine,mxda,m-phenylenebis methylamine,methylamine, m-phenylenebis
15133
[3-(aminomethyl)phenyl]methanamine
C1=CC(=CC(=C1)CN)CN
Density 1.0300g/mL
Refractive Index 1.5700 to 1.572
Boiling Point 265°C (745.0mmHg)
Flash Point >112°C
Specific Gravity 1.03
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 100mL
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H4(CH2NH2)2
Beilstein 13, IV, 293
Formula Weight 136.2
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material m-Xylylenediamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger