Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Morphine-D3-3-beta-D-glucuronide Solution, 100 μg/mL in Methanol with 0.05% NaOH, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco M0171ML
Description
Morphine-3β-D-glucuronide is a primary plasma and urinary metabolite of the opiate analgesic morphine. Sold under trade names including MS Contin™, Kadian™, Oramorph™, and Roxanol for pain relief, morphine is also a commonly abused recreational drug. This stable-labeled internal standard is suitable for quantitation of morphine levels in urine, serum, or plasma by LC/MS or GC/MS for pain prescription monitoring, urine drug testing, clinical toxicology, or forensic analysis.Specifications
67-56-1, 136765-44-1 | |
MFCD01075376 | |
100 μg/mL in methanol with 0.05% NaOH | |
Ampule | |
11°C (51.8°F) - closed cup |
C23H24NO9D3 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
464.48 | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : 2 to 8°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction