Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Teledyne SSI M1 Class Pump
The compact SSI M1 Class HPLC Pump consists of single-headed, positive displacement piston pumps that deliver high-performance and precision.
$2470.00 - $2590.00
Specifications
For Use With (Application) Dispensing, Metering, Separation
View More Specs
Products 4
Catalog Number Mfr. No. Flow Rate Pressure Material Includes Price Quantity & Availability  
Catalog Number Mfr. No. Flow Rate Pressure Material Includes Price Quantity & Availability  
06-657-001 View Documents Teledyne SSI
M1010PFN3A
10 mL/min 2000 PSI Polyether Ether Ketone (PEEK) Flouropolymer Energizer Standard Piston Seal Each for $2,470.00
N/A
06-657-002 View Documents Teledyne SSI
M1010SFN1A
10 mL/min 2000 PSI Stainless Steel Stainless Steel Energizer Standard Piston Seal Each for $2,590.00
N/A
06-657-003 View Documents Teledyne SSI
M1040PFN3A
40 mL/min 500 PSI Polyether Ether Ketone (PEEK) Flouropolymer Energizer Standard Piston Seal Each for $2,470.00
N/A
06-657-004 View Documents Teledyne SSI
M1040SFN1A
40 mL/min 500 PSI Stainless Steel Stainless Steel Energizer Standard Piston Seal Each for $2,590.00
N/A
Description
Description
  • Self-Flush and Piston Wash
  • No Pulse Dampener or Pressure & Leak Sensor
  • Inlet Tubing: 1/8in. OD Flexible
  • Outlet Tubing: 1/16in. OD
  • Applications include precise chemical metering, dispensing, and specific HPLC separations. Standard fluid path materials are Stainless Steel and PEEK. With 10 mL/min and 40 mL/min versions, reaching pressures up to 2,000 PSI, these pumps capture the majority of typical laboratory fluid transfer applications.
  • Features include interactive keypad control, Pulse Dampener and Outlet Filter, RS-232 Serial and Micro USB 2.0 ports for complete PC control and status, as well as hardware for laboratory stand mounting. Having the smallest possible footprint for a high pressure pump, the M1 Class will easily integrate into any laboratory or industrial environment.
Specifications
Specifications
Dispensing, Metering, Separation