Learn More
Swift™ M10T-HD Series Advanced Microscope with Integrated HD Camera Products for Science Education
Ideal for higher education and professional applications.
$1946.00 - $2226.00
Specifications
Condenser Type 1.25 Abbe, Rack and Pinion, Iris
Eyepiece W10XD, 20mm
Head Style Trinocular
Lighting Type Variable, Corded, 3w LED
View More Specs
Products 2
Catalog Number Mfr. No. Model Magnification Power Price Quantity & Availability  
Catalog Number Mfr. No. Model Magnification Power Price Quantity & Availability  
S43544 Products for Science Education
View Documents
Swift™
M10THDP
M10T-HD-P 4XD, 10XD, 40XRD, 100XRD Plan Each for $1,946.00
Add to cart
S43543 Products for Science Education
View Documents
Swift™
M10THDS
M10T-HD-S 4XD, 10XD, 40XRD, 100 XRD Semi- Plan Each for $2,226.00
Add to cart
Description
Description
Available with semi-plan, plan or multi-phase objectives lenses
HD Camera Specifications:
  • HDMI output: 1080P
  • 1920 X 1980 captured images and video
  • 60 FPS (Full HD)
  • On-board software for control and capture
  • SD card slot (32 GB max)
  • USB mouse support
Specifications
Specifications
1.25 Abbe, Rack and Pinion, Iris
W10XD, 20mm
Trinocular
Variable, Corded, 3w LED