Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M13BacteriophageProteing3p Mouse anti-Virus, Clone: E1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266601

Catalog No. NBP266601


Add to cart

Description

Description

M13BacteriophageProteing3p Monoclonal antibody specifically detects M13BacteriophageProteing3p in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications

Specifications

M13BacteriophageProteing3p
E1
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500
Purified
Hybridoma produced by the fusion of splenocytes from mice immunized with isolated M13 phage coat proteins and mouse myeloma cells.
Protein A or G purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PBS with 0.08% Sodium Azide
Mouse
IgG2a
100μg
Primary
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit