Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

M13BacteriophageProteing8p Mouse anti-Virus, Clone: RL-ph2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266602

Catalog No. NBP266602


Add to cart

Description

Description

M13BacteriophageProteing8p Monoclonal antibody specifically detects M13BacteriophageProteing8p in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

M13BacteriophageProteing8p
RL-ph2
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
PBS with 0.09% Sodium Azide
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with isolated M13 phage coat proteins and mouse SP2/0-Ag14 myeloma cells.
Monoclonal
Protein G purified
Store at -20°C. Avoid freeze-thaw cycles.
ELISA,Flow Cytometry,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unconjugated
Purified
Mouse
IgG2a
Primary
100μg
Virus
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit