Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Methoxetamine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M1561ML
Description
A certified solution standard for use in LC/MS or GC/MS applications in clinical toxicology, forensic analysis, or urine drug testing. Methoxetamine, with street names such as MXE, Mexxy, and Roflcoptr, is a designer drug and ketamine derivative used recreationally as a dissociative and hallucinogen. The drug has been reported to have the same unwanted side effects as ketamine. Methoxetamine use is reportedly popular in the United Kingdom with cases also observed in the US.Specifications
67-56-1,1239908-48-5 | |
9.7°C (49.5°F) | |
C15H21NO2 · HCl | |
Ampule | |
-20°C |
1.0 mg/mL in methanol (as free base) | |
283.79 | |
MFCD27966836 | |
1 mL | |
UN 1230 - class 3 - PG 2 - Methanol, solution |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation