Learn More
CERILLIANT™ Metronidazole Solution, 2.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M1831ML
Description
Metronidazole, sold under trade names such as Flagyl™, Filmet, Rozex, MetroGel™, and Anabact, is a nitroimidazole antibiotic drug used in the treatment and prevention of bacteria and protozoa infections. This Certified Spiking Solution™ is suitable as starting material for calibrators, controls, or linearity standards for therapeutic drug monitoring or clinical and diagnostic testing of metronidazole in patient blood, serum, or plasma samples by LC-MS/MS, LC or GC/MS.Specifications
67-56-1,443-48-1 | |
9.7°C (49.5°F) - closed cup | |
C6H9N3O3 | |
Ampule | |
-20°C |
2.0 mg/mL in methanol | |
171.15 | |
MFCD00009750 | |
1 mL | |
UN 1230 - class 3 - PG 2 - Methanol |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation