Learn More
CERILLIANT™ Maprotiline-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco M2031ML
Catalog No. 01-001-676
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This stable-labeled internal standard is suitable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS. Maprotiline is a tetracyclic antidepressant marketed under the trade name Ludiomil for treatment of depression.
Specifications
Specifications
67-56-1
9.7°C (49.5°F) - closed cup
C20D3H20N · HCl
1 mL
100 μg/mL in methanol (as free base)
316.88
Ampule
-20°C
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation