Learn More
CERILLIANT™ para-Methoxybutyryl fentanyl-13C6 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco M2130.5ML
Catalog No. 01-001-683
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
para-Methoxybutyryl fentanyl, also known as 4-methoxybutyryl fentanyl, is a fentanyl analog and sold on the illicit market as a designer drug. This stable-labeled internal standard is suitable for quantification of para-methoxybutyryl fentanyl levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.
Specifications
Specifications
67-56-1
422.94
Ampule
-20°C
100 μg/mL in methanol (as free base)
C1813C6H32N2O2 · HCl
0.5 mL
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation