Learn More
Swift M3601C-USB Compound Microscope Digital Camera Package
Features cordless, energy-efficient LED illumination and a student-friendly design.
Manufacturer: Swift M3601CUSB
Description
Camera Features:
- Model: D-Moticam 2
- Live, digital output
- Detachable USB cable
- Feature-packed Motic Images Plus software
Specifications
Monocular | |
Camera: 2.0 MP (1600 X 1200) | |
Built-in N.A. 0.65 with iris |
Variable, Cordless, LED | |
M3601C | |
4X, 10XD, 40XRD Achromat |