Learn More
Tocris Bioscience™ MA 2029 Available on GSA/VA Contract for Federal Government customers only.
Potent and selective, competitive motilin receptor antagonist; orally active
Manufacturer:  Tocris Bioscience™ 4934/10
 View more versions of this product
Catalog No. 49-341-0
Add to cart
This item is not returnable. View return policy
Description
Description
Potent and selective, competitive motilin receptor antagonist (IC50 = 4.9 nM). Selective for the motilin receptor over a range of other receptors and ion channels. Inhibits motilin-induced duodenal muscle contractions in vitro. Also inhibits motilin-induced colonic and abdominal contractions in vivo. Orally active.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
MA 2029
>98
10mg
MA 2029
556.71