Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MA 2029
Potent and selective, competitive motilin receptor antagonist; orally active
$210.00 - $881.00
Specifications
Product Type | MA 2029 |
---|---|
Inhibitors | MA 2029 |
Molecular Weight | 556.71 |
Purity | >98 |
Description
Potent and selective, competitive motilin receptor antagonist (IC50 = 4.9 nM). Selective for the motilin receptor over a range of other receptors and ion channels. Inhibits motilin-induced duodenal muscle contractions in vitro. Also inhibits motilin-induced colonic and abdominal contractions in vivo. Orally active.Specifications
MA 2029 | |
MA 2029 | |
556.71 | |
>98 |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title