Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAB21L3 Mouse anti-Human, Clone: OTI1F6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525507

Catalog No. PIMA525507


Add to cart

Description

Description

MAB21L3 is a protein coding gene.
Specifications

Specifications

MAB21L3
Monoclonal
0.75 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N8X9
C1orf161
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
126868
Flow Cytometry, Western Blot
OTI1F6
Unconjugated
MAB21L3
Liquid
MAB21L3
Full length human recombit protein of C1orf161 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit