Learn More
Thermo Scientific™ MAbPac™ RP LC Analytical Columns PROMO
Provides higher resolution using reversed-phase conditions. MAbPac RP LC Analytical Columns PROMO separates and characterizes monoclonal antibodies using HPLC and LC/MS. Receive promotional offer of 20%
Manufacturer:  Thermo Scientific™ 088647PM
 View more versions of this product
Catalog No. 08-864-7PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over
Designed for reversed-phase biomolecule LC and LC/MS separations
  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput
Specifications
Specifications
Analytical
100 mm
DVB
0 to 14
<110°C
2.1 mm
4000psi
4 µm
1500 Å