Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns

Use these SEC columns for monoclonal antibody (mAb) analysis, including the high-resolution separation of monomers, aggregates and fragments.

$600.00 - $2110.00

Specifications

Particle Size 5 µm
Packing Material Spherical, Fully Porous, Ultrapure Silica
pH 2.5 to 7.5
Stationary Phase Diol (OH)
Temperature 30°C
View More Specs

Products 5
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Column Type Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Column Type Price Quantity & Availability  
13-622-467
View Documents
Thermo Scientific™
088460
5 µm 7.8 mm 300 mm Size Exclusion
Each for $2,110.00
Add to cart
 
DX074696
View Documents
Thermo Scientific™
074696
5 µm 4 mm 300 mm Size Exclusion
Each for $1,798.00
Add to cart
 
DX074697
View Documents
Thermo Scientific™
074697
5 µm 4 mm 50 mm Size Exclusion
Each for $600.00
Add to cart
 
DX075592
View Documents
Thermo Scientific™
075592
5 µm 4 mm 150 mm Size Exclusion
Each for $1,366.00
Add to cart
 
03-898-145
View Documents
Thermo Scientific™
088790
5 µm 2.1 mm 150 mm Size Exclusion
Each for $1,289.00
Add to cart
 
Description

Description

Use Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns for high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis. These size-exclusion chromatography (SEC) columns provide superior performance, even under nondenaturing conditions, and in high- and low-salt mobile phases and volatile eluents.

High-Performance SEC Columns for mAb Analysis
  • Hydrophilic bonded layer minimizes undesired interactions between the biomolecules and the stationary phase
  • Nonmetallic and biocompatible PEEK housing eliminates metal contamination from the column hardware
  • Stable surface bonding leads to low column bleed and compatibility with MS, ELSD and Thermo Scientific™ Dionex™ Corona™ CAD detection
  • Rugged column packing provides reliability, reproducibility and long column life
  • Columns are available in 5µm particle size and 150 and 300mm lengths, with an average pore diameter of 300Å
Specifications

Specifications

5 µm
2.5 to 7.5
30°C
Spherical, Fully Porous, Ultrapure Silica
Diol (OH)
SDS

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit