Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Machine/Equipment Label, Header: DANGER/PELIGRO, Legend: HIGH VOLTAGE……ONLY/ALTO VOLTAJE
Communicates important safety information and machine operational instructions. Brady Machine and Equipment Labels are made of durable, self-sticking polyester and most feature pictograms as well as text to inform safety procedures and potential hazards with machine operation.
Supplier: Brady™ 86235
Description
- Placed on machine or equipment to increase safety
- Made of B-302 self sticking polyester
- Available in size (W x H): 8.89 x 12.7cm (3.5 x 5 in.)
Specifications
Acrylic, Permanent, Cold Temperature | |
Gloss | |
Machine/Equipment Label | |
English, Spanish | |
Rectangle | |
88.9 x 127 mm (W x H) |
Black, Red, White | |
Self-adhesive Polyester (B-302) | |
Header: DANGER/PELIGRO, Legend: HIGH VOLTAGE AUTHORIZED PERSONNEL ONLY/ALTO VOLTAJE | |
5/Pk. | |
3.5 x 5 in. (W x H) | |
0.01 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction