Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Machine/Equipment Label, Header: REMEMBER, Legend: LOCK, TAG, CLEAR, TRY

Communicates important safety information and machine operational instructions. Brady Machine and Equipment Labels are made of durable, self-sticking polyester and most feature pictograms as well as text to inform safety procedures and potential hazards with machine operation.

Manufacturer:  Brady™ 86250

Catalog No. 19-110-690


Add to cart

Description

Description

  • Placed on machine or equipment to increase safety
  • Made of B-302 self sticking polyester
  • Available in size (W x H): 12.7 x 8.89cm (5 x 3.5 in.)
Specifications

Specifications

Acrylic, Permanent, Cold temperature
Header: REMEMBER, Legend: LOCK, TAG, CLEAR, TRY
Rectangle
W x H: 127 x 88.9mm
English
Machine/Equipment Label
Black, Red, White
Gloss
W x H: 5 x 3.5 in.
0.01 in.
Self Sticking Polyester (B-302)
5/pk
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit