Learn More
Mature Macrophage Marker Mouse anti-Human, Clone: HAM56, eBioscience™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen 14654893
Catalog No. 50-112-2709
Add to cart
Description
Description
Description: The HAM56 monoclonal antibody recognizes human macrophages. This includes tingible macrophages, interdigitating macrophages of lymph nodes, and tissue macrophages such as Kupffer cells of the liver and alveolar macrophages of the lung. HAM56 also recognizes monocytes as well as endothelial cells of capillaries and blood vessels. HAM56 does not react with lymphocytes, platelets, granulocytes, or red blood cells. Applications Reported: This HAM56 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunohistochemical staining. Applications Tested: This HAM56 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval. This can be used at less than or equal to 1:2000. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.
Specifications
Specifications
Mature Macrophage Marker
Monoclonal
Unconjugated
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Immunohistochemistry (Paraffin)
HAM56
tris with 1% BSA and 0.09% sodium azide; pH 7.3-7.7
ED2-like antigen; Mature Macrophages; Macrophages
IgM, kappa
100 μL
4° C
Primary
Human
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only.