Learn More
Macrophage Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0029-11H3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212436AF350
Catalog No. NP212436A30
Add to cart
Description
Description
Macrophage Monoclonal antibody specifically detects Macrophage in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Macrophage
RM0029-11H3
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase
Isolated mouse macrophages
0.1 mL
Primary
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Rat
IgG2
Store at 4°C in the dark.
Monoclonal