Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Macrophage Rat anti-Mouse, DyLight 350, Clone: RM0029-11H3, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212436UV0.1ML
Description
Macrophage Monoclonal specifically detects Macrophage in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Macrophage | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rat | |
>95% pure by SDS-PAGE | |
RUO | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0029-11H3 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
Isolated mouse macrophages | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction