Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Mad Max™ Thermo Waterproof Synthetic Leather Gloves with TPR Padding

Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are lined with 3M™ Thinsulate™ for thermal protection, without the bulkiness of fleece. The synthetic leather palm enhances grip in in dry, or light oil applications.

  • Synthetic leather palm provides dexterity, and enhances grip
  • PVC sandy grip on palm and fingertips provides additional cut/abrasion resistance
  • 3M™ Thinsulate™ lining provides thermal protection without the bulk of fleece
  • HIPORA™ liner is waterproof, windproof, and breathable
  • Thermoplastic Rubber (TPR) molded finger, thumb, and back hand guards for maximum protection
  • Reinforced thumb crotch for extended glove life, enhanced comfort, and increased abrasion resistance
  • Airprene wrist is made with breathable neoprene for comfort
  • Synthetic leather upper, and heavy Lycra™ fabric bottom fourchettes
  • Hi-vis coloring and reflective back
Cuff Material Nylon Airprene
Color Hi-Vis Green, Black
Hand Specific
Material Synthetic Leather
Product Type Mechanics Gloves
Size Large
For Use With (Application) Construction, Auto Repair, Manufacturing, and Landscaping
Glove Cuff Color Black
Glove Cuff Style Open
Liner Material 3M™ Thinsulate™
Lining HIPORA™
Series 120-4070
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less