Learn More
MAD2L1 Mouse anti-Human, DyLight 680, Clone: 1761, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243820FR
Catalog No. NBP243820FR
Add to cart
Description
Description
MAD2L1 Monoclonal antibody specifically detects MAD2L1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
MAD2L1
1761
Western Blot
Purified
MAD2L1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human MAD2L1. The exact sequence is proprietary.
Protein G purified
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary
4085
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
HSMAD2, MAD2 (mitotic arrest deficient, yeast, homolog)-like 1, MAD2 mitotic arrest deficient-like 1 (yeast), MAD2-like protein 1, MAD2mitotic arrest deficient, yeast, homolog-like 1, Mitotic arrest deficient 2-like protein 1, mitotic spindle assembly checkpoint protein MAD2A
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human