Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287766
Description
MAD3 Polyclonal specifically detects MAD3 in Human samples. It is validated for Western Blot.Specifications
MAD3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
83463 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
bHLHc13, Class C basic helix-loop-helix protein 13, MAD3FLJ35523, MAX dimerization protein 3, Max dimerizer 3, Max-associated protein 3, Max-interacting transcriptional repressor MAD3, MGC2383, MYX | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human MAD3 | |
100 μL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction